Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTTCCTCCCCAGTACGTGGGAG[C/T]CCTAGAGGACATGTTGCAGGCCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602129 MIM: 610675 | ||||||||||||||||||||
Literature Links: |
MYO9B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYO9B - myosin IXB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USE1 - unconventional SNARE in the ER 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018467.3 | 173 | Missense Mutation | GCC,GTC | A,V 38 | NP_060937.1 | |
XM_017026975.1 | 173 | Missense Mutation | GCC,GTC | A,V 38 | XP_016882464.1 | |
XM_017026976.1 | 173 | Intron | XP_016882465.1 |