Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATAAATTTCCTGTGACCTTGTTA[C/T]GGGAAAATAATAAGCCCCTGCTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607261 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EVC2 - EvC ciliary complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166136.1 | 3952 | Intron | NP_001159608.1 | |||
NM_147127.4 | 3952 | Intron | NP_667338.3 | |||
XM_017007735.1 | 3952 | UTR 3 | XP_016863224.1 | |||
XM_017007736.1 | 3952 | Intron | XP_016863225.1 | |||
XM_017007737.1 | 3952 | Intron | XP_016863226.1 | |||
XM_017007738.1 | 3952 | Intron | XP_016863227.1 | |||
XM_017007739.1 | 3952 | Intron | XP_016863228.1 |
LINC01587 - long intergenic non-protein coding RNA 1587 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |