Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGATGGGGAAAAGTTAAGGAAAGT[G/T]CAGGTGAGAATGGGCAGAGAGAAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS48 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRSS48 - protease, serine 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183375.2 | 924 | Missense Mutation | GCA,TCA | A,S 160 | NP_899231.2 | |
XM_011531921.2 | 924 | Missense Mutation | GCA,TCA | A,S 183 | XP_011530223.1 | |
XM_011531922.2 | 924 | Intron | XP_011530224.1 |
Set Membership: |
HapMap |