Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTCCTGACTGTTATCACGATCT[C/T]TCTTCCTCACATTGTCCTAGTAACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYNPO2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SYNPO2 - synaptopodin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128933.2 | Intron | NP_001122405.1 | ||||
NM_001128934.2 | Intron | NP_001122406.1 | ||||
NM_001286754.1 | Intron | NP_001273683.1 | ||||
NM_001286755.1 | Intron | NP_001273684.1 | ||||
NM_133477.2 | Intron | NP_597734.2 |
Set Membership: |
HapMap |