Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTGGTATAAGGAACCTAACCAA[C/T]TTATTTACGTGGCTTTCTGAAAAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609132 MIM: 601422 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM1A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KDM1A - lysine demethylase 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LUZP1 - leucine zipper protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142546.1 | 4794 | UTR 3 | NP_001136018.1 | |||
NM_033631.3 | 4794 | UTR 3 | NP_361013.3 | |||
XM_011542090.2 | 4794 | UTR 3 | XP_011540392.1 | |||
XM_011542091.2 | 4794 | UTR 3 | XP_011540393.1 | |||
XM_017002252.1 | 4794 | UTR 3 | XP_016857741.1 |