Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACAGGTTACACCATAAATCAAT[A/T]TCACAGTTCAGTTTTATAATCTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614843 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ODAM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ODAM - odontogenic, ameloblast asssociated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017855.3 | Intron | NP_060325.3 | ||||
XM_005265698.3 | Intron | XP_005265755.1 | ||||
XM_006714245.3 | Intron | XP_006714308.1 | ||||
XM_017008331.1 | Intron | XP_016863820.1 |