Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATGTGAAAGTAAAACATTTTGA[C/T]GATAATAACTGAAACCAAAATAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606701 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRGX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DRGX - dorsal root ganglia homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276451.1 | 2277 | Intron | NP_001263380.1 | |||
XM_011540089.2 | 2277 | UTR 3 | XP_011538391.1 |
Set Membership: |
HapMap |