Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCACATCTATTAATGTTTAAAA[C/G]TAATGTCCCTTGGCCAGGCGCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600722 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CAP1 - adenylate cyclase associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPT1 - palmitoyl-protein thioesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000310.3 | Intron | NP_000301.1 | ||||
NM_001142604.1 | Intron | NP_001136076.1 | ||||
XM_005271008.1 | Intron | XP_005271065.1 |
Set Membership: |
HapMap |