Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGACTCATCAGGGACACCAAGGC[A/G]CAAAACATGTCATTCTGTTCCAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610043 MIM: 614683 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL23A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL23A1 - collagen type XXIII alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173465.3 | Intron | NP_775736.2 | ||||
XM_006714933.3 | Intron | XP_006714996.1 | ||||
XM_006714934.3 | Intron | XP_006714997.1 | ||||
XM_006714935.3 | Intron | XP_006714998.1 | ||||
XM_006714936.3 | Intron | XP_006714999.1 | ||||
XM_011534688.2 | Intron | XP_011532990.1 | ||||
XM_011534689.2 | Intron | XP_011532991.1 | ||||
XM_011534690.2 | Intron | XP_011532992.1 | ||||
XM_011534691.2 | Intron | XP_011532993.1 | ||||
XM_011534692.2 | Intron | XP_011532994.1 | ||||
XM_017010018.1 | Intron | XP_016865507.1 |
PHYKPL - 5-phosphohydroxy-L-lysine phospho-lyase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |