Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTAGTTAAACTGGCAGTTGTAA[C/T]TTGGACTTGTAAAATGAAATTAACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612275 MIM: 610275 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGNBP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
GGNBP2 - gametogenetin binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024835.4 | Intron | NP_079111.1 | ||||
XM_005257689.2 | Intron | XP_005257746.1 | ||||
XM_011525265.2 | Intron | XP_011523567.1 | ||||
XM_011525266.1 | Intron | XP_011523568.1 | ||||
XM_017025107.1 | Intron | XP_016880596.1 |
PIGW - phosphatidylinositol glycan anchor biosynthesis class W | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |