Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACACCGGGAATTTAATGAGTGA[A/C]GTAACACTGAAAGCACATGATGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605922 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMPA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IMPA2 - inositol monophosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014214.2 | Intron | NP_055029.1 | ||||
XM_011525659.2 | Intron | XP_011523961.1 | ||||
XM_011525661.2 | Intron | XP_011523963.1 |
Set Membership: |
HapMap |