Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTAACTGTACTACCTGTGACCT[G/T]TATGCTTTGTGCAAAGTCTGTCTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613482 MIM: 611833 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCNL2 - cyclin L2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC148413 - uncharacterized LOC148413 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL20 - mitochondrial ribosomal protein L20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318485.1 | Intron | NP_001305414.1 | ||||
NM_017971.3 | Intron | NP_060441.2 |
Set Membership: |
HapMap |