Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGTCCAGGCTGGTGCTTGTTTT[A/T]TTAAGCTGGTCAAGGACATTTAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603631 MIM: 608754 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RPS9 - ribosomal protein S9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013.3 | Intron | NP_001004.2 | ||||
NM_001321701.1 | Intron | NP_001308630.1 | ||||
NM_001321702.1 | Intron | NP_001308631.1 | ||||
NM_001321704.1 | Intron | NP_001308633.1 | ||||
NM_001321705.1 | Intron | NP_001308634.1 | ||||
NM_001321706.1 | Intron | NP_001308635.1 | ||||
XM_017027112.1 | Intron | XP_016882601.1 |
TSEN34 - tRNA splicing endonuclease subunit 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |