Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTACATCATTCTGTTCTCACCTC[A/G]ATTAGCCAAGACCTGCTGTTTCTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605514 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDH15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PCDH15 - protocadherin related 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142763.1 | 8209 | Intron | NP_001136235.1 | |||
NM_001142764.1 | 8209 | Intron | NP_001136236.1 | |||
NM_001142765.1 | 8209 | Intron | NP_001136237.1 | |||
NM_001142766.1 | 8209 | Intron | NP_001136238.1 | |||
NM_001142767.1 | 8209 | Intron | NP_001136239.1 | |||
NM_001142768.1 | 8209 | Intron | NP_001136240.1 | |||
NM_001142769.1 | 8209 | Intron | NP_001136241.1 | |||
NM_001142770.1 | 8209 | Intron | NP_001136242.1 | |||
NM_001142771.1 | 8209 | UTR 3 | NP_001136243.1 | |||
NM_001142772.1 | 8209 | UTR 3 | NP_001136244.1 | |||
NM_001142773.1 | 8209 | Intron | NP_001136245.1 | |||
NM_033056.3 | 8209 | Intron | NP_149045.3 | |||
XM_017016571.1 | 8209 | Intron | XP_016872060.1 | |||
XM_017016572.1 | 8209 | UTR 3 | XP_016872061.1 | |||
XM_017016573.1 | 8209 | Intron | XP_016872062.1 |
Set Membership: |
HapMap |