Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATACAAAAAATAAAAAGACCTAG[G/T]CTGTACCCTTAAGCTGAAGTAACAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRCC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LRRCC1 - leucine rich repeat and coiled-coil centrosomal protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033402.4 | Intron | NP_208325.3 | ||||
XM_005251318.3 | Intron | XP_005251375.1 | ||||
XM_005251319.4 | Intron | XP_005251376.1 | ||||
XM_005251320.3 | Intron | XP_005251377.1 | ||||
XM_005251322.3 | Intron | XP_005251379.1 | ||||
XM_017013920.1 | Intron | XP_016869409.1 | ||||
XM_017013921.1 | Intron | XP_016869410.1 | ||||
XM_017013922.1 | Intron | XP_016869411.1 | ||||
XM_017013923.1 | Intron | XP_016869412.1 | ||||
XM_017013924.1 | Intron | XP_016869413.1 | ||||
XM_017013925.1 | Intron | XP_016869414.1 | ||||
XM_017013926.1 | Intron | XP_016869415.1 | ||||
XM_017013927.1 | Intron | XP_016869416.1 | ||||
XM_017013928.1 | Intron | XP_016869417.1 |