Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATGAGGAGGAGGAGGAGGAGGA[C/G]GAAGCCTTCGAGGATGAGGGAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610303 | ||||||||||||||||||||
Literature Links: |
MAFA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAFA - MAF bZIP transcription factor A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZC3H3 - zinc finger CCCH-type containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015117.2 | 2648 | Silent Mutation | TCC,TCG | S,S 873 | NP_055932.2 | |
XM_006716536.3 | 2648 | Silent Mutation | TCC,TCG | S,S 935 | XP_006716599.2 | |
XM_011516943.2 | 2648 | Intron | XP_011515245.1 | |||
XM_011516944.2 | 2648 | Intron | XP_011515246.1 | |||
XM_017013248.1 | 2648 | Silent Mutation | TCC,TCG | S,S 1001 | XP_016868737.1 | |
XM_017013249.1 | 2648 | Silent Mutation | TCC,TCG | S,S 1001 | XP_016868738.1 |