Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGCAGAGGAGGGACAATGGCC[C/T]GGCTCCTGGAGGCAAGTGTTGGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107266 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD22 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD22 - CD22 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185099.1 | Intron | NP_001172028.1 | ||||
NM_001185100.1 | Intron | NP_001172029.1 | ||||
NM_001185101.1 | Intron | NP_001172030.1 | ||||
NM_001278417.1 | Intron | NP_001265346.1 | ||||
NM_001771.3 | Intron | NP_001762.2 |