Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTAAATAACATTGACATACAGAG[A/C]AGTTAGGTGATTTGTCTGAGACCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605197 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KYNU PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KYNU - kynureninase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032998.1 | Intron | NP_001028170.1 | ||||
NM_001199241.1 | Intron | NP_001186170.1 | ||||
NM_003937.2 | Intron | NP_003928.1 | ||||
XM_011512102.2 | Intron | XP_011510404.1 | ||||
XM_017005217.1 | Intron | XP_016860706.1 |
Set Membership: |
HapMap |