Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAGCTTTAATCATAAGTAGGGT[A/G]ATTGGTAGAACTCCTTTCCTCCTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605683 MIM: 613334 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAZ2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BAZ2B - bromodomain adjacent to zinc finger domain 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MARCH7 - membrane associated ring-CH-type finger 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282805.1 | Intron | NP_001269734.1 | ||||
NM_001282806.1 | Intron | NP_001269735.1 | ||||
NM_001282807.1 | Intron | NP_001269736.1 | ||||
NM_022826.3 | Intron | NP_073737.1 | ||||
XM_005246773.2 | Intron | XP_005246830.1 | ||||
XM_005246774.1 | Intron | XP_005246831.1 | ||||
XM_017004718.1 | Intron | XP_016860207.1 | ||||
XM_017004719.1 | Intron | XP_016860208.1 | ||||
XM_017004720.1 | Intron | XP_016860209.1 | ||||
XM_017004721.1 | Intron | XP_016860210.1 |
Set Membership: |
HapMap |