Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCCTGTGTTCTTGGCATGACCA[T/C]GAGAGAAGCGTGCTCGCTCTGGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180290 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RBP3 - retinol binding protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF488 - zinc finger protein 488 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153034.2 | 1936 | UTR 3 | NP_694579.1 | |||
XM_006717617.3 | 1936 | UTR 3 | XP_006717680.1 | |||
XM_011539244.2 | 1936 | UTR 3 | XP_011537546.1 | |||
XM_017015642.1 | 1936 | UTR 3 | XP_016871131.1 | |||
XM_017015643.1 | 1936 | UTR 3 | XP_016871132.1 |