Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGAATCTGACTTTACAGATGATA[A/T]CTCGAGCTGGGTTTACCAGCTAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601237 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF112 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RNF112 - ring finger protein 112 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007148.4 | 202 | Intron | NP_009079.2 | |||
XM_006721571.3 | 202 | UTR 5 | XP_006721634.1 | |||
XM_006721572.3 | 202 | UTR 5 | XP_006721635.1 | |||
XM_006721573.3 | 202 | Intron | XP_006721636.1 |
Set Membership: |
HapMap |