Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTGAACTTTCTTAGACATCAGC[C/G]CAGAAACGGAGATGGAGGCTGTGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604705 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MERTK PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MERTK - MER proto-oncogene, tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006343.2 | Intron | NP_006334.2 | ||||
XM_005263565.4 | Intron | XP_005263622.1 | ||||
XM_005263568.4 | Intron | XP_005263625.1 | ||||
XM_011510490.2 | Intron | XP_011508792.1 | ||||
XM_017003164.1 | Intron | XP_016858653.1 | ||||
XM_017003165.1 | Intron | XP_016858654.1 |
Set Membership: |
HapMap |