Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCAGGACTTCCTGTAATTACTTA[C/T]GGCCCCTCACAATACCCTCACGGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613358 MIM: 611480 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH16A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ALDH16A1 - aldehyde dehydrogenase 16 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145396.1 | Intron | NP_001138868.1 | ||||
NM_153329.3 | Intron | NP_699160.2 | ||||
XM_011526441.1 | Intron | XP_011524743.1 | ||||
XM_011526442.1 | Intron | XP_011524744.1 |
PIH1D1 - PIH1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |