Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTAAGGCAGCAGAAGCACAATGC[A/G]GAGGGAGCATCCGAACTGCCTGGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611186 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf61 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C1orf61 - chromosome 1 open reading frame 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320453.1 | Intron | NP_001307382.1 | ||||
NM_001320454.1 | Intron | NP_001307383.1 | ||||
NM_001320455.1 | Intron | NP_001307384.1 | ||||
NM_006365.2 | Intron | NP_006356.1 | ||||
XM_011509067.1 | Intron | XP_011507369.1 | ||||
XM_011509068.1 | Intron | XP_011507370.1 | ||||
XM_011509070.1 | Intron | XP_011507372.1 | ||||
XM_011509071.2 | Intron | XP_011507373.1 | ||||
XM_011509072.1 | Intron | XP_011507374.1 | ||||
XM_011509073.2 | Intron | XP_011507375.1 | ||||
XM_011509074.1 | Intron | XP_011507376.1 | ||||
XM_017000061.1 | Intron | XP_016855550.1 | ||||
XM_017000062.1 | Intron | XP_016855551.1 | ||||
XM_017000063.1 | Intron | XP_016855552.1 | ||||
XM_017000064.1 | Intron | XP_016855553.1 | ||||
XM_017000065.1 | Intron | XP_016855554.1 | ||||
XM_017000066.1 | Intron | XP_016855555.1 |
MIR9-1 - microRNA 9-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |