Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCTGTGGAGAAAACAGCACAAT[C/G]ATATCTCTTCTCATTCTTTAGTCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616472 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf189 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C1orf189 - chromosome 1 open reading frame 189 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1orf43 - chromosome 1 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098616.2 | Intron | NP_001092086.1 | ||||
NM_001297717.1 | Intron | NP_001284646.1 | ||||
NM_001297718.1 | Intron | NP_001284647.1 | ||||
NM_001297720.1 | Intron | NP_001284649.1 | ||||
NM_001297721.1 | Intron | NP_001284650.1 | ||||
NM_001297723.1 | Intron | NP_001284652.1 | ||||
NM_015449.3 | Intron | NP_056264.1 | ||||
NM_138740.3 | Intron | NP_620077.1 |
UBAP2L - ubiquitin associated protein 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |