Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCATACAAATTCCATATCCAGT[A/G]AAAATCATTTTGATCCACAATCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614451 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELOVL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ELOVL7 - ELOVL fatty acid elongase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104558.1 | 3836 | UTR 3 | NP_001098028.1 | |||
NM_001297617.1 | 3836 | UTR 3 | NP_001284546.1 | |||
NM_001297618.1 | 3836 | UTR 3 | NP_001284547.1 | |||
NM_024930.2 | 3836 | UTR 3 | NP_079206.2 | |||
XM_005248606.4 | 3836 | UTR 3 | XP_005248663.1 | |||
XM_005248607.3 | 3836 | UTR 3 | XP_005248664.1 | |||
XM_006714695.3 | 3836 | UTR 3 | XP_006714758.1 | |||
XM_011543651.2 | 3836 | UTR 3 | XP_011541953.1 | |||
XM_017009885.1 | 3836 | UTR 3 | XP_016865374.1 | |||
XM_017009886.1 | 3836 | UTR 3 | XP_016865375.1 | |||
XM_017009887.1 | 3836 | UTR 3 | XP_016865376.1 | |||
XM_017009888.1 | 3836 | Intron | XP_016865377.1 | |||
XM_017009889.1 | 3836 | Intron | XP_016865378.1 |
Set Membership: |
HapMap |