Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAAAGAGTTTATTTTACGCTCCT[C/T]TTTCCCTGCCTTCTCCTCCACATTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM267 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM267 - transmembrane protein 267 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022483.4 | Intron | NP_071928.2 | ||||
XM_005248333.4 | Intron | XP_005248390.1 | ||||
XM_005248335.3 | Intron | XP_005248392.1 | ||||
XM_006714486.3 | Intron | XP_006714549.1 | ||||
XM_011514074.2 | Intron | XP_011512376.1 | ||||
XM_011514075.2 | Intron | XP_011512377.1 |