Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTCCATCTGTGAAATGGGACAA[C/T]AGATGTTCAAGTCTAGCCCTGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603346 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPAS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NPAS1 - neuronal PAS domain protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321086.1 | Intron | NP_001308015.1 | ||||
NM_002517.3 | Intron | NP_002508.2 | ||||
XM_006723231.2 | Intron | XP_006723294.1 | ||||
XM_011526997.1 | Intron | XP_011525299.1 | ||||
XM_011526999.1 | Intron | XP_011525301.1 | ||||
XM_011527001.1 | Intron | XP_011525303.1 | ||||
XM_011527002.1 | Intron | XP_011525304.1 | ||||
XM_011527006.1 | Intron | XP_011525308.1 | ||||
XM_011527007.1 | Intron | XP_011525309.1 | ||||
XM_017026841.1 | Intron | XP_016882330.1 | ||||
XM_017026842.1 | Intron | XP_016882331.1 | ||||
XM_017026843.1 | Intron | XP_016882332.1 | ||||
XM_017026844.1 | Intron | XP_016882333.1 |
Set Membership: |
HapMap |