Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATATTGACTGAACTCTGTAGATG[A/G]GCCCTGAGTGTGGTTTTTCAGAGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604691 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AKAP6 - A-kinase anchoring protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004274.4 | Intron | NP_004265.3 | ||||
XM_011537378.2 | Intron | XP_011535680.1 | ||||
XM_011537379.2 | Intron | XP_011535681.1 | ||||
XM_011537383.2 | Intron | XP_011535685.1 | ||||
XM_017021808.1 | Intron | XP_016877297.1 | ||||
XM_017021809.1 | Intron | XP_016877298.1 | ||||
XM_017021810.1 | Intron | XP_016877299.1 | ||||
XM_017021811.1 | Intron | XP_016877300.1 |