Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609774 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
API5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
API5 - apoptosis inhibitor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142930.1 | Intron | NP_001136402.1 | ||||
NM_001142931.1 | Intron | NP_001136403.1 | ||||
NM_001243747.1 | Intron | NP_001230676.1 | ||||
NM_006595.3 | Intron | NP_006586.1 | ||||
XM_006718359.3 | Intron | XP_006718422.1 | ||||
XM_017018464.1 | Intron | XP_016873953.1 | ||||
XM_017018465.1 | Intron | XP_016873954.1 |