Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCATGAAAATATCTGGAGAAAG[C/T]ACAGGGCGATTTATGGGAGTTTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607580 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC22A10 - solute carrier family 22 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039752.3 | Intron | NP_001034841.3 | ||||
XM_011545016.2 | Intron | XP_011543318.1 | ||||
XM_017017699.1 | Intron | XP_016873188.1 | ||||
XM_017017700.1 | Intron | XP_016873189.1 | ||||
XM_017017701.1 | Intron | XP_016873190.1 | ||||
XM_017017702.1 | Intron | XP_016873191.1 | ||||
XM_017017703.1 | Intron | XP_016873192.1 | ||||
XM_017017704.1 | Intron | XP_016873193.1 |
Set Membership: |
HapMap |