Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604932 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKIG PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PKIG - protein kinase (cAMP-dependent, catalytic) inhibitor gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281444.1 | Intron | NP_001268373.1 | ||||
NM_001281445.1 | Intron | NP_001268374.1 | ||||
NM_007066.4 | Intron | NP_008997.1 | ||||
NM_181804.2 | Intron | NP_861520.1 | ||||
NM_181805.2 | Intron | NP_861521.1 | ||||
XM_017027614.1 | Intron | XP_016883103.1 | ||||
XM_017027615.1 | Intron | XP_016883104.1 | ||||
XM_017027616.1 | Intron | XP_016883105.1 |
Set Membership: |
HapMap |