Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCTCTGCTTTCTAGCCTTTCTAG[A/G]CCTGAACATCCTGCCTCCTGCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609031 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPPIN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPPIN - epididymal peptidase inhibitor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EPPIN-WFDC6 - EPPIN-WFDC6 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198986.1 | 929 | UTR 3 | NP_001185915.1 |
WFDC6 - WAP four-disulfide core domain 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080827.1 | 929 | Intron | NP_543017.1 |