Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTTTAGGTGTTATCAAAATTAT[C/T]GTCACATTTTTCAGGTTTGTAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF680 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF680 - zinc finger protein 680 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130022.1 | 1746 | Intron | NP_001123494.1 | |||
NM_178558.4 | 1746 | Missense Mutation | AAT,GAT | N,D 525 | NP_848653.2 | |
XM_006715959.2 | 1746 | Intron | XP_006716022.1 | |||
XM_017012126.1 | 1746 | Intron | XP_016867615.1 | |||
XM_017012127.1 | 1746 | Intron | XP_016867616.1 | |||
XM_017012128.1 | 1746 | Intron | XP_016867617.1 | |||
XM_017012129.1 | 1746 | UTR 3 | XP_016867618.1 |