Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGGATACAGGTGAATGACCCCA[A/G]GAATCAATCAGATATGGGGTCATTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 613536 | |||||||||||||||||||||||
Literature Links: |
LEKR1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LEKR1 - leucine, glutamate and lysine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004316.2 | Intron | NP_001004316.2 | ||||
NM_001193283.1 | Intron | NP_001180212.1 | ||||
XM_005247441.3 | Intron | XP_005247498.1 | ||||
XM_005247442.3 | Intron | XP_005247499.1 | ||||
XM_006713631.3 | Intron | XP_006713694.1 | ||||
XM_006713636.2 | Intron | XP_006713699.1 | ||||
XM_011512803.2 | Intron | XP_011511105.1 | ||||
XM_011512804.2 | Intron | XP_011511106.1 | ||||
XM_011512806.2 | Intron | XP_011511108.1 | ||||
XM_011512807.2 | Intron | XP_011511109.1 | ||||
XM_011512808.2 | Intron | XP_011511110.1 | ||||
XM_011512809.1 | Intron | XP_011511111.1 | ||||
XM_011512811.2 | Intron | XP_011511113.1 | ||||
XM_011512813.2 | Intron | XP_011511115.1 | ||||
XM_011512814.2 | Intron | XP_011511116.1 | ||||
XM_011512815.2 | Intron | XP_011511117.1 | ||||
XM_011512816.2 | Intron | XP_011511118.1 | ||||
XM_017006374.1 | Intron | XP_016861863.1 | ||||
XM_017006375.1 | Intron | XP_016861864.1 |