Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGCGATTCGTGGGGCCCTTAGG[C/T]GTGCTGGTGAGAGTCACAAATTGAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605799 MIM: 614062 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMN PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AMN - amnion associated transmembrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030943.3 | Intron | NP_112205.2 |
CDC42BPB - CDC42 binding protein kinase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006035.3 | Intron | NP_006026.3 | ||||
XM_005268227.1 | Intron | XP_005268284.1 | ||||
XM_005268228.1 | Intron | XP_005268285.1 | ||||
XM_005268229.1 | Intron | XP_005268286.1 | ||||
XM_005268230.4 | Intron | XP_005268287.1 | ||||
XM_011537387.1 | Intron | XP_011535689.1 |
Set Membership: |
HapMap |