Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATCTCCGTGGAGACCAAGCGGT[C/T]GAGCGGCGCCCGCTCTGCAGCCTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612270 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDCA4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDCA4 - cell division cycle associated 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017955.3 | 373 | Missense Mutation | AAC,GAC | N,D 93 | NP_060425.2 | |
NM_145701.2 | 373 | Missense Mutation | AAC,GAC | N,D 93 | NP_663747.1 |
Set Membership: |
HapMap |