Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTATTTCACCTGCTGATGTTA[C/T]CCAGGTTCCCGTTTATTTTCATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616918 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIGG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIGG - phosphatidylinositol glycan anchor biosynthesis class G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127178.2 | Intron | NP_001120650.1 | ||||
NM_001289051.1 | Intron | NP_001275980.1 | ||||
NM_001289052.1 | Intron | NP_001275981.1 | ||||
NM_001289053.1 | Intron | NP_001275982.1 | ||||
NM_001289055.1 | Intron | NP_001275984.1 | ||||
NM_001289057.1 | Intron | NP_001275986.1 | ||||
NM_017733.4 | Intron | NP_060203.3 | ||||
XM_005272284.2 | Intron | XP_005272341.1 | ||||
XM_011513490.2 | Intron | XP_011511792.1 | ||||
XM_011513491.1 | Intron | XP_011511793.1 | ||||
XM_011513492.1 | Intron | XP_011511794.1 | ||||
XM_011513493.1 | Intron | XP_011511795.1 | ||||
XM_011513494.2 | Intron | XP_011511796.1 | ||||
XM_017008321.1 | Intron | XP_016863810.1 | ||||
XM_017008322.1 | Intron | XP_016863811.1 | ||||
XM_017008323.1 | Intron | XP_016863812.1 | ||||
XM_017008324.1 | Intron | XP_016863813.1 | ||||
XM_017008325.1 | Intron | XP_016863814.1 | ||||
XM_017008326.1 | Intron | XP_016863815.1 |
ZNF721 - zinc finger protein 721 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |