Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605317 | ||||||||||||||||||||
Literature Links: |
FOXP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOXP2 - forkhead box P2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172766.2 | 248 | Intron | NP_001166237.1 | |||
NM_001172767.2 | 248 | Intron | NP_001166238.1 | |||
NM_014491.3 | 248 | Intron | NP_055306.1 | |||
NM_148898.3 | 248 | Intron | NP_683696.2 | |||
NM_148899.3 | 248 | Intron | NP_683697.2 | |||
NM_148900.3 | 248 | Intron | NP_683698.2 | |||
XM_017012801.1 | 248 | UTR 5 | XP_016868290.1 |