Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTAAAAATGAGTATGTGCTGCCT[C/T]TCAGGGTTATCTGTTGAGTTCTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607985 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf89 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C16orf89 - chromosome 16 open reading frame 89 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098514.2 | Intron | NP_001091984.2 | ||||
NM_152459.4 | Intron | NP_689672.4 | ||||
XM_005255143.3 | Intron | XP_005255200.2 | ||||
XM_011522392.2 | Intron | XP_011520694.1 | ||||
XM_017022972.1 | Intron | XP_016878461.1 | ||||
XM_017022973.1 | Intron | XP_016878462.1 | ||||
XM_017022974.1 | Intron | XP_016878463.1 |
NAGPA - N-acetylglucosamine-1-phosphodiester alpha-N-acetylglucosaminidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAGPA-AS1 - NAGPA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |