Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTCTCTTTGTGTTCCTAGGCCC[A/G]CAAAGTCTTTGGTCTGAAGACTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612872 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NKAIN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NKAIN3 - Na+/K+ transporting ATPase interacting 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304533.1 | Intron | NP_001291462.1 | ||||
XM_011517511.2 | Intron | XP_011515813.1 | ||||
XM_011517512.2 | Intron | XP_011515814.1 | ||||
XM_017013359.1 | Intron | XP_016868848.1 | ||||
XM_017013360.1 | Intron | XP_016868849.1 | ||||
XM_017013361.1 | Intron | XP_016868850.1 |
UG0898H09 - uncharacterized LOC643763 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |