Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGTGGCTTATAACCGAGAGAAA[C/T]TGAAGTCTACTTGCCAGTGTGTGGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605966 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNF4G PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HNF4G - hepatocyte nuclear factor 4 gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004133.4 | Intron | NP_004124.4 | ||||
XM_011517516.2 | Intron | XP_011515818.1 | ||||
XM_017013373.1 | Intron | XP_016868862.1 | ||||
XM_017013374.1 | Intron | XP_016868863.1 | ||||
XM_017013375.1 | Intron | XP_016868864.1 | ||||
XM_017013376.1 | Intron | XP_016868865.1 |
Set Membership: |
HapMap |