Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACAAGAAAAAGATATGAATTTGG[C/T]AATGGATATGTCTAAGAGGTGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606940 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFHX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZFHX4 - zinc finger homeobox 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024721.4 | Intron | NP_078997.4 | ||||
XM_011517592.2 | Intron | XP_011515894.1 | ||||
XM_011517593.2 | Intron | XP_011515895.1 | ||||
XM_011517594.2 | Intron | XP_011515896.1 | ||||
XM_011517595.2 | Intron | XP_011515897.1 | ||||
XM_011517596.2 | Intron | XP_011515898.1 | ||||
XM_011517597.2 | Intron | XP_011515899.1 | ||||
XM_017013845.1 | Intron | XP_016869334.1 |
ZFHX4-AS1 - ZFHX4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |