Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCGGTGTCGATGGCGGTGGTGA[G/A]GTGGTGGAAGAGTGGCTTGCTGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615168 MIM: 604394 | ||||||||||||||||||||
Literature Links: |
AMZ1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMZ1 - archaelysin family metallopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284355.2 | 1200 | Intron | NP_001271284.1 | |||
NM_001321766.1 | 1200 | Intron | NP_001308695.1 | |||
NM_133463.3 | 1200 | Intron | NP_597720.1 | |||
XM_011515150.2 | 1200 | Intron | XP_011513452.1 | |||
XM_011515151.2 | 1200 | Intron | XP_011513453.1 | |||
XM_017011774.1 | 1200 | Intron | XP_016867263.1 | |||
XM_017011775.1 | 1200 | Intron | XP_016867264.1 | |||
XM_017011776.1 | 1200 | Intron | XP_016867265.1 |
GNA12 - G protein subunit alpha 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282440.1 | 1200 | Missense Mutation | CTC,TTC | L,F 274 | NP_001269369.1 | |
NM_001282441.1 | 1200 | Missense Mutation | CTC,TTC | L,F 291 | NP_001269370.1 | |
NM_001293092.1 | 1200 | Missense Mutation | CTC,TTC | L,F 333 | NP_001280021.1 | |
NM_007353.2 | 1200 | Missense Mutation | CTC,TTC | L,F 350 | NP_031379.2 | |
XM_011515288.2 | 1200 | Missense Mutation | CTC,TTC | L,F 253 | XP_011513590.1 | |
XM_017012008.1 | 1200 | Missense Mutation | CTC,TTC | L,F 222 | XP_016867497.1 |