Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGTCTGCCCAAAAAAGTAGCT[A/C]AAGAAACATTAAAAAAGAAAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602838 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3C2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIK3C2B - phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002646.3 | Intron | NP_002637.3 | ||||
XM_005245257.2 | Intron | XP_005245314.1 | ||||
XM_005245258.3 | Intron | XP_005245315.1 | ||||
XM_011509630.2 | Intron | XP_011507932.1 | ||||
XM_011509631.2 | Intron | XP_011507933.1 | ||||
XM_011509633.2 | Intron | XP_011507935.1 | ||||
XM_017001473.1 | Intron | XP_016856962.1 | ||||
XM_017001474.1 | Intron | XP_016856963.1 | ||||
XM_017001475.1 | Intron | XP_016856964.1 |
Set Membership: |
HapMap |