Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACTCTGGTAACTCATTGCCAGT[C/G]TGAATTTTCAATAGGCCAGATGGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606109 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENY2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ENY2 - ENY2, transcription and export complex 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193557.1 | Intron | NP_001180486.1 | ||||
NM_020189.5 | Intron | NP_064574.1 |
NUDCD1 - NudC domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128211.1 | Intron | NP_001121683.1 | ||||
NM_032869.3 | Intron | NP_116258.2 | ||||
XM_017013910.1 | Intron | XP_016869399.1 | ||||
XM_017013911.1 | Intron | XP_016869400.1 |
Set Membership: |
HapMap |