Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTATCTTGTTCCAGTTTTTTGG[A/G]AAAGGCATTTATGTGAGGTATATAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615667 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC6L2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ERCC6L2 - ERCC excision repair 6 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010895.2 | Intron | NP_001010895.1 | ||||
NM_020207.4 | Intron | NP_064592.2 | ||||
XM_011518641.2 | Intron | XP_011516943.1 | ||||
XM_011518644.2 | Intron | XP_011516946.1 | ||||
XM_011518645.2 | Intron | XP_011516947.1 | ||||
XM_011518646.2 | Intron | XP_011516948.1 | ||||
XM_011518647.1 | Intron | XP_011516949.1 | ||||
XM_011518648.2 | Intron | XP_011516950.1 | ||||
XM_011518650.2 | Intron | XP_011516952.1 | ||||
XM_017014707.1 | Intron | XP_016870196.1 |
LINC00476 - long intergenic non-protein coding RNA 476 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |