Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTTACTGTGGTACAGAAATGGA[A/G]AATGGGTTTGAGAGAAATAGGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMIM14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SMIM14 - small integral membrane protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317896.1 | Intron | NP_001304825.1 | ||||
NM_001317897.1 | Intron | NP_001304826.1 | ||||
NM_174921.2 | Intron | NP_777581.1 | ||||
XM_011513640.2 | Intron | XP_011511942.1 | ||||
XM_017007862.1 | Intron | XP_016863351.1 | ||||
XM_017007863.1 | Intron | XP_016863352.1 |
UGDH-AS1 - UGDH antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |