Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATGAGCAAGAGGGTCAGCTTC[C/G]ATTTTAGCTGGTCTGAGTTGGCGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PHF14 - PHD finger protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014660.3 | Intron | NP_055475.2 | ||||
XM_017012811.1 | Intron | XP_016868300.1 | ||||
XM_017012812.1 | Intron | XP_016868301.1 | ||||
XM_017012813.1 | Intron | XP_016868302.1 | ||||
XM_017012814.1 | Intron | XP_016868303.1 | ||||
XM_017012815.1 | Intron | XP_016868304.1 | ||||
XM_017012816.1 | Intron | XP_016868305.1 | ||||
XM_017012817.1 | Intron | XP_016868306.1 |
Set Membership: |
HapMap |