Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAACATCGATCTGAAGGATTCAT[A/G]AGGAGCTTATCTGAACAGATTTATC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603390 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE8B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
WDR41 - WD repeat domain 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018268.3 | 2954 | Intron | NP_060738.2 | |||
XM_005248551.4 | 2954 | Intron | XP_005248608.1 | |||
XM_005248552.3 | 2954 | Intron | XP_005248609.1 | |||
XM_011543505.2 | 2954 | Intron | XP_011541807.1 | |||
XM_011543507.2 | 2954 | Intron | XP_011541809.1 | |||
XM_017009606.1 | 2954 | Intron | XP_016865095.1 |
Set Membership: |
HapMap |